Would you like to log in again?
Whatman™ UNIPLATE™ Collection and Analysis Microplates
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
UNIPLATE™ Whatman™ UNIPLATE™
7701-1651 7701-5110 7701-3250 7701-3250 7701-5110 7701-5102 7701-1651 7701-5200 7701-5102 7701-5500 7701-5500 7701-5200
13503-188PK 501.91 USD InStock
CA28317-472 CA13503-188 13503-188 28317-472 13503-204 CA13503-152 13503-170 CA13503-170 13503-190 13503-152 CA13503-204 CA13503-190
Whatman™ UNIPLATE™ Collection and Analysis Microplates
Microplates HTS Microplates
The UNIPLATE™ series includes microplates with a range of well profiles, well volumes and well densities, made from diverse polymer materials. Most UNIPLATE™ microplates conform to the proposed SBS microplate standard and fit most microplate readers and automated plate handling devices.
They are suitable for a wide range of applications, including simple filtrate collection, when used in conjunction with UNIFILTER® microplates, as well as homogeneous assay techniques utilised in HTS.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty