Would you like to log in again?
PYREX® Griffin Beakers, Graduated, Corning
Supplier: Corning
PYREX®
1000-600 1000-250 1000-10 1000-PACK 1000-4L 1000-1L 1000-150 1000-1L 1000-100 1000-600 1000-2L 1000-4L 1000-3L 1000-400 1000-800 1000-250 1000-50 1000-400 1000-50 1000-100 1000-30 1000-20
470211-328CS 765.8 USD InStock
470211-328 89090-522 13912-149 13912-127 13912-207 13912-229 13912-240 13912-284 13912-262 89090-524 13912-069 13912-320 13912-091 470211-314 470211-326 470211-330 13912-182 13912-160 470211-320 470211-332 470211-310 89091-542
PYREX® Griffin Beakers, Graduated, Corning
Beakers
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • Thermal shock resistance
Manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength.
Sizes 250 ml and larger have a double graduated metric scale to indicate approximate content. The 10 ml size is not graduated. All sizes have an extra large marking spot.
Certifications: Meet ASTM E960 specifications.
Ordering information: The variety pack includes one of each of the following beakers: 50, 100, 250, 600 and 1000 ml.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty