Would you like to log in again?
PYREX® Heavy-Duty Griffin Beakers, Graduated
Supplier: Corning
PYREX®
1003-150 1003-600 1003-4L 1003-1L 1003-2L 1003-400 1003-250
89090-528CS 449.3 USD InStock
89090-528 89090-526 13912-502 13912-568 13912-546 13912-524 13912-604
PYREX® Heavy-Duty Griffin Beakers, Graduated
Beakers
PYREX® Heavy-Duty Griffin beakers are made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • Thermal shock resistance
  • With extra large marking spot
Heavy-duty beakers are specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. PYREX® Heavy-Duty Griffin beakers in sizes 250 ml and larger have a double graduated metric scale to indicate approximate content.
Certifications: Meet ASTM E960 specifications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty