Would you like to log in again?
VWR®, Beaker Cups, Polystyrene
Supplier: VWR
VWR®
B150-100 B50-500 B10-1000 B5-1000 B20-500 B250-100
13916-037CS 147.13 USD InStock
13916-037 13916-015 13916-004 13915-996 13915-985 13916-060
VWR®, Beaker Cups, Polystyrene
Beakers
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
  • Disposable
  • Polystyrene fabrication
  • Economical sample cup
The 5 mL and 20 mL sizes are recommended for flame photometry analysis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty