Would you like to log in again?
Chemware® Beakers, Teflon® PFA, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
Chemware®
D1069536 D1069539 D1069537 D1069538
13917-586EA 342.12 USD InStock
13917-586 13917-584 13917-582 13917-580
Chemware® Beakers, Teflon® PFA, Saint-Gobain Performance Plastics
Beakers
These low metal ion content PFA beakers are ideal for trace metal analysis and ultra-pure applications.
  • Special machined bottom design for rapid heat transfer
  • Transparent for easy viewing
  • Graduated for easy measuring
  • Non-stick, easy to clean surface
Microwave heatable, or heat directly on hot plate up to 500°F (260°C) when proper care is exercised.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty