Would you like to log in again?
ORION® Conductivity Standards, Thermo Scientific
Supplier: Thermo Scientific
Orion™
01100610 01100710 01100510
14002-866PK 74.06 USD InStock
14002-866 14002-864 14002-862
ORION® Conductivity Standards, Thermo Scientific
Standards Conductivity Standards
These ready-to-use standards allow five conductivity levels to be easily measured in the field
Standards come in convenient 15mL pouches and certified against standards whose calibrations are traceable to the National Institute of Standards and Technology.
Ordering information: Store at room temperature.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty