Would you like to log in again?
Super Lube White Grease, Food Grade, R.S. Hughes
Supplier: R.S. Hughes
21030
14003-020CS 324.2 USD InStock
14003-020
Super Lube White Grease, Food Grade, R.S. Hughes
Lubricants
Super lube white grease is comprised of PTFE and synthetic used for automotive equipment applications.
  • Easy to use
  • Clean, non-toxic, non-staining
  • Dielectric
  • USDA Rating H-1
  • Will not drip, run or evaporate
  • Impervious to salt water, safe in potable water
  • Long lasting
  • Prevents rust and corrosion while reducing friction
  • Wide Temperature Range -45°to 450°F
Certifications: Meets NSF H1 standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty