Would you like to log in again?
Simultaneous Start Timer, Sper Scientific
Supplier: Sper Scientific
810027 810027C
14003-166EA 17.6 USD InStock
14003-166 14003-168
Simultaneous Start Timer, Sper Scientific
Timers Digital Timers
This simultaneous start multi-channel timer is unique in two respects; it is the only multi-channel timer on which various channels can be set independently but started simultaneously.
  • Hourly chime
  • Unit beeps every hour
It also has a loud alarm, which is strong enough to be heard above high levels of background noise. 4 channels count either up or down to 100 hours with 1-second resolution. A 5th channel works as a calendar and alarm clock. A 6th channel is a 10 hour stopwatch with ¹/₁₀₀ second. resolution, lap and split functions. Has a built in stand for bench top use, clips to a pocket, or sticks to any metal surface with a magnetic back.
Certifications: N.I.S.T. traceable certificate of compliance for 14003-168.
Ordering information: Two button cell batteries included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty