Would you like to log in again?
HYPO-CHLOR® Cleaning Agent, Veltek Associates
Supplier: Veltek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HYPO-CHLOR®
SHC-13Z-5.25 SHC-16Z-0.52 SHC-02-0.25 SHC-02-5.25 SHC-16Z-0.25 SHC-02-0.52 SHC-01-0.52 SHC-01-5.25 SHC-16Z-5.25
14003-328CS 706.38 USD InStock
14003-328 14003-336 14003-324 14218-554 75795-370 14003-334 14218-556 14003-333 14218-552
HYPO-CHLOR® Cleaning Agent, Veltek Associates
Cleaning Agents Detergents and Surface Cleansers
HYPO-CHLOR® is an effective one step, ready-to-use product that is available in premixed concentrations of 5.25%, 0.52%, and 0.25%.
  • Filled in ISO 5 (Grade A/B, Former Class 100)
  • Made with USP Water for Injection
  • Filtered at 0.2 microns
  • Sterility tested according to current USP compendium
  • Assayed according to current USP compendium
  • Completely traceable from start to finish
  • Completely validated for sterility and shelf life
Sodium hypochlorite solutions, formulated with WFI, used to clean cleanrooms and controlled areas including those used in health care institutions, biopharmaceutical, medical device, and diagnostic manufacturing facilities. HYPO-CHLOR® Products were developed for use in cleaning rotation cycles for maintaining a clean environment
Sterile sodium hypochlorite solutions are filtered at 0.2µm, gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation
Product uses: Use on hard, non-porous, and inanimate surfaces, on materials such as plastic, glass, vinyl, glazed porcelain, and stainless steel. Can also be used on machinery, tools, tables, counters, laminar flow benches, floors, walls and carts.
Delivery information: The 473 ml (16 oz.) sizes come with sterile spray attachments.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty