Would you like to log in again?
Process QDS® Disinfectant Cleaner, STERIS®
Supplier: Steris
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1J7501 1J7508
21899-730CS 365.16 USD InStock
21899-730 14004-114
Process QDS® Disinfectant Cleaner, STERIS®
Cleaning Agents Disinfectants
Process QDS® Process and Research Cleaner is a phosphate-free, one-step cleaner/disinfectant formulated for use on hard non-porous surfaces. This product is recommended for use in disinfecting, sanitizing, and deodorizing hard non-porous surfaces. It is effective in hard water and organic soil loads.
  • Efficacy - Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy.
  • Productivity - Formulation includes surfactants for surface cleaning while disinfecting.
  • Reliability - Ideal for use in a contamination control rotation.
  • Flexibility - Can be used as a disinfectant or sanitizer.
  • Assurance - Complete lot traceability with each lot documented by Certificate of Analysis.
The use-dilution can be applied manually or sprayed.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty