Would you like to log in again?
Cage-Klenz® 200 Acid-Based Cage Wash Detergent, Steris
Supplier: Steris
Cage-Klenz®
1K2005 1K2002 1K2005 1K2002
14004-124EA 2047.28 USD InStock
CA14004-124 CA14223-964 14004-124 14223-964
Cage-Klenz® 200 Acid-Based Cage Wash Detergent, Steris
Cleaning Agents Detergents and Surface Cleansers
High-performance, phosphoric acid-based detergent system removes hard water scale, urine scale, metal oxide, and a wide spectrum of soils
Free-rinsing detergent minimizes residue. Low-foaming and suitable for use in high-energy automatic washing applications. pH (1%): 2.0 (typical).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty