Would you like to log in again?
Cage-Klenz® 220 Acid-Based Detergents
Supplier: Steris
Cage-Klenz®
1K22D3 1K2205 1K2202
77531-228EA 9374.85 USD InStock
77531-228 14004-182 14004-184
Cage-Klenz® 220 Acid-Based Detergents
Cleaning Agents Detergents and Surface Cleansers
Phosphate-free, hydroxyacetic acid-based liquid detergent removes hard water scale, urine scale, organic soils, and metal oxides
Manufactured in an FDA-registered and ISO 9001/13485-certified facility.
Free-rinsing detergent minimizes residue. Low-foaming and suitable for use in high-energy automatic washing applications. pH (1%): 2.8 (typical).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty