Would you like to log in again?
CIP 150® Chlorinated Alkaline Cleaner, STERIS®
Supplier: Steris
CIP 150®
1D1505 1D1505
14004-232EA 430 USD InStock
14004-232 CA14004-232
CIP 150® Chlorinated Alkaline Cleaner, STERIS®
Cleaning Agents Detergents and Surface Cleansers
Formulated to serve as an effective validatable cleaner for pharmaceutical, biotechnology, cosmetic, nutraceutical, and other industries
Combines effective detergency with oxidizing agents to rapidly clean oily, proteinaecous soils. Low foaming at all temperatures and can be used in both CIP and COP applications. Has both specific and non-specific methods to detect cleaning agent residues, including HPLC, TOC, conductivity, and others. Free rinsing. Complete lot traceability. Easily removed from product surfaces
Certifications: Manufactured within an FDA and ISO 9001 registered facility.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty