Would you like to log in again?
Orion™ pH Electrode Filling Solution, Thermo Scientific
Supplier: Thermo Scientific
Orion™
900011 900002 610011 810007 910008-WA 900004 900019 900003 510011 900001
34108-019PK 192.14 USD InStock
34108-019 34108-008 34111-850 34107-413 14004-258 34108-020 34107-806 34111-830 34107-990 14231-146
Orion™ pH Electrode Filling Solution, Thermo Scientific
Electrolytes Electrode Filling Solutions
Use Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes.
Only use ROSS filling solution with ROSS™ and ROSS Ultra™ pH electrodes to prevent damage. When the single junction reference electrode or double junction reference electrode are used with Orion™ ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty