Would you like to log in again?
Orion™ Calcium Electrodes, Thermo Scientific
Supplier: Thermo Scientific
ORION® ionplus® Sure-Flow® ORION®
9720BNWP 972001 932001 9320BN
34109-576EA 476.95 USD InStock
34109-576 34116-618 34118-118 14004-262
Orion™ Calcium Electrodes, Thermo Scientific
Electrodes ISE Electrodes
Measure calcium in aqueous solutions simply, accurately, and economically with Orion™ Calcium Electrodes.
  • Free-flowing Sure-Flow junction assures stable, drift-free potentials
  • A separate reference electrode is unnecessary, making this electrode convenient to use with small samples
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty