This tape is designed specifically for sealing wafer boxes and cleanroom packaging and bag sealing.
- No residues after prolonged storage
- Easy release from the roll with low static charge
Tape adheres lightly to plastics, stretches easily around corners, and has low metal content.
Delivery information: Supplied clean and double-bagged.