Would you like to log in again?
DEC-QUAT® 100, Veltek Associates
Supplier: Veltek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DECON-QUAT®
DQ100-03-8Z DQ100-06-16Z-01 DQ100-04-2Z DQ100-05-2Z DQ100-01 DQ100-07-16Z-01 DQ100-03-8Z DQ100-02
75795-368CS 540.34 USD InStock
75795-368 14003-702 14004-490 14004-488 14003-701 14004-492 14003-706 BA14004-492KPH
DEC-QUAT® 100, Veltek Associates
Cleaning Agents Disinfectants
DEC-QUAT® 100 is a quaternary ammonium solution for use on hard non-porous environmental surfaces.
  • Effective as a broad spectrum hospital disinfectant, fungicide, deodorizer, virucide
  • Food and non-food contact sanitizer effective against a broad spectrum of pathogenic bacteria
  • Available as both sterile and non-sterile
  • Completely traceable and has been validated for sterility and shelf life
This product is designed for use in hospital, medical and health care institutions as a broad-spectrum disinfectant solution on hard, non-porous environmental surfaces such as walls, ceilings, floors, and countertops. Effective against Pseudomonas aeruginosa, Staphylococcus aureus, and Salmonella choleraesuis. Effective against Hepatitis B Virus when contact time is 10 minutes at 1 oz:1 gal of water. Effective against Human Immunodeficiency Virus Type 1 (HIV) after 30 seconds of contact time.
Packaging: Filled sterile by filtration at 0.2 microns. Gamma-irradiated, double-bagged and delivered with lot-specific sterile documentation.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty