Would you like to log in again?
Automation Pipette Tips for Tecan Te-MO and Freedom EVO Workstations, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
162-96R 163-96RS 163-96R 161-96RS 907-262 907-261 162-96RS 906-021 LMF163-96RS-50
14004-624CS 527.35 USD InStock
14004-624 14004-613 14004-626 14004-620 72830-590 72830-586 14004-628 14004-618 72830-588
Automation Pipette Tips for Tecan Te-MO and Freedom EVO Workstations, Thermo Scientific
Pipette Tips
Thermo Scientific provides a solution for your Tecan robotic pipette tips.
These automation pipette tips have been designed, validated, and tested on a Tecan EVO robot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our Tecan workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty