Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sterile Scalpel Blades, Walter Stern
Supplier: Walter Stern
320-003 320-001 320-005 320-002
25608-065PK 106.81 USD InStock
25608-065 25608-087 14006-940 14006-938
Sterile Scalpel Blades, Walter Stern
Scalpels and Scalpel Assemblies Scalpel Blades
Fit handles 25607-925 and 25607-947
Made of high-grade vacuum hardened steel with excellent cutting ability. Sterilized by gamma radiation. Packed individually in aluminum foil.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty