Would you like to log in again?
Sample Vials with Writing Patch, Glass, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences
225012 225014
14007-176CS 198.56 USD InStock
14007-176 66011-372
Sample Vials with Writing Patch, Glass, Wheaton, DWK Life Sciences
Vials
Wheaton Sample Vials are made from low extractable borosilicate glass to provide superior chemical resistance.
  • Autoclavable vials and caps
Vials feature a large writing patch and have attached caps to maintain cleanliness. Caps include a 14B rubber liner that is suitable for aqueous samples.
Ordering information: Vials conform to ASTM E438 Type 1, Class A and USP Type 1 standards. Packaged in Lab File® partitioned trays that provide an easy way to inventory samples or store empty vials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty