Would you like to log in again?
SP Wilmad-LabGlass Auto Liquid Dividing Distilling Heads, SP Industries
Supplier: SP Industries
LG-6171-102 LG-6171-100
14008-408EA 3612.29 USD InStock
14008-408 14008-410
SP Wilmad-LabGlass Auto Liquid Dividing Distilling Heads, SP Industries
Distillation Components Distillation Heads
These borosilicate glass distilling heads have an interchangeable Standard Taper inner joint at the bottom for connection to a distilling column and a full length Standard Taper 10/30 outer member to receive a thermometer calibrated for 75mm immersion.
  • A tilting funnel in the head receives the condensate and normally returns the liquid to the column
  • A glass-enclosed iron core attached to the funnel may be energized by an external solenoid at timed intervals to tip the funnel and deliver the liquid instead to a product receiver
  • The takeoff tube has a Socket Joint 18/9 ball
  • The head is completely vacuum jacketed
  • The jacket has an internal bellows to allow for differential expansion of jacket and head and is silvered to improve accuracy of temperature measurements and minimize heat transfer
Other combinations of interchangeable joint sizes - Standard Taper, Socket Joint and pipe flange - can be supplied. For a solenoid to use with these heads see LG-6215; for product receivers see LG-6190.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty