Would you like to log in again?
SP Wilmad-LabGlass Lattice Clamps, SP Industries
Supplier: SP Industries
LG-4622-100
14200-050EA 23.11 USD InStock
14200-050
SP Wilmad-LabGlass Lattice Clamps, SP Industries
Clamps General Purpose Clamps
These stamped steel clamps are zinc plated for corrosion resistance and have a steel thumbscrew.
Clamps are designed for use in lattice systems, and fit rod sizes up to 3/4" diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty