Would you like to log in again?
SP Wilmad-LabGlass Solid Glass Beads, SP Industries
Supplier: SP Industries
LG-6750-106 LG-6750-104 LG-6750-102
14200-198EA 270.19 USD InStock
14200-198 14200-636 14200-250
SP Wilmad-LabGlass Solid Glass Beads, SP Industries
Beads Glass Beads
These borosilicate glass beads are used for distilling column packing.
They vary in size and are sold in 1 lb. packages.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty