Would you like to log in again?
SP Wilmad-LabGlass Calibration/Measuring Flask Set Supplements, TC Metric System, SP Industries
Supplier: SP Industries
LG-4319-112
14200-518EA 630.04 USD InStock
14200-518
SP Wilmad-LabGlass Calibration/Measuring Flask Set Supplements, TC Metric System, SP Industries
Flasks Volumetric Flasks
These borosilicate glass graduated neck flasks come in sizes to supplement our official metric series, LG-4317
Additional sizes can be supplied on request. Calibrated "to contain" at 68°F/20°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty