Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Wilmad-LabGlass Calibration/Measuring Flask Set, TC Metric System, SP Industries

Supplier: SP Industries
LG-4317-108 LG-4317-100 LG-4317-104 LG-4317-112 LG-4317-102 LG-4317-110 LG-4317-106
14200-712EA 871.84 USD InStock
14200-712 14201-078 14200-736 14201-220 14212-738 14201-086 14200-580
SP Wilmad-LabGlass Calibration/Measuring Flask Set, TC Metric System, SP Industries
Flasks Volumetric Flasks
Borosilicate glass cylinder and flasks are calibrated to contain (TC) at 68°F/20°C.

  • Sizes included are 50, 100, 250, 500, 1000, and 2000 mL
  • The 50 mL is cylindrical, graduated 10 to 50 mL in 1 mL divisions
  • All others are graduated neck flasks, graduated above and below the nominal

Ordering information: The complete set includes a case which holds all six sizes.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty