Would you like to log in again?
SP Wilmad-LabGlass Calibration/Liquid Measuring Flasks, TC U.S. Series, SP Industries
Supplier: SP Industries
LG-4310-100
14200-958EA 7121.89 USD InStock
14200-958
SP Wilmad-LabGlass Calibration/Liquid Measuring Flasks, TC U.S. Series, SP Industries
Flasks Volumetric Flasks
Intended for checking automatic fill volumes, the borosilicate glass cylinder and flasks are calibrated "to contain" (TC) at 68°F
The complete set includes two cases – one with a 2 oz. graduated cylinder and larger sized flasks consisting of 1 gill, 1/2 pint, 1 pint, 1 quart, ½ gal. The 1 gal flask comes in a case of its own.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty