Would you like to log in again?
SP Wilmad-LabGlass Capillary Tip Centrifuge Tube, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LG-4474-100
14201-246EA 159.98 USD InStock
14201-246
SP Wilmad-LabGlass Capillary Tip Centrifuge Tube, SP Industries
Tubes Centrifuge Tubes
Designed for use in ASTM D-2273 and ASTM D-2709, this borosilicate glass capillary tipped long form centrifuge tube is 100 ml in length.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty