Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SP Wilmad-LabGlass Round Bottom Short Single Neck Flasks, SP Industries
Supplier: SP Industries
LG-7220-202 LG-7220-148 LG-7220-156 ML-1110-718 ML-1110-728 ML-1110-774 ML-1110-762 LG-7220-154 ML-1110-714 ML-1110-710 LG-7220-142 LG-7220-246 LG-7220-158 ML-1110-708 ML-1110-768 LG-7220-248 LG-7220-146 ML-1110-732 ML-1110-772 ML-1110-778 LG-7220-152 LG-7220-160 ML-1110-722 LG-7220-270 LG-7220-150 LG-7220-242 LG-7220-140
14202-080EA 67.21 USD InStock
14202-080 14202-082 14201-956 14211-284 14201-910 14211-464 14211-028 14209-468 14209-550 14202-088 14202-144 14202-124 14202-084 14210-920 14202-090 14201-660 14202-778 14209-466 14206-196 14206-494 14210-054 14211-290 14202-310 14211-030 14211-470 14201-618 14201-718
SP Wilmad-LabGlass Round Bottom Short Single Neck Flasks, SP Industries
Flasks Round Bottom Flasks
With excellent chemical durability for contamination-free results, these short neck, borosilicate glass, round bottom boiling flasks have a single neck with a full-length Standard Taper outer joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty