Would you like to log in again?
SP Wilmad-LabGlass Air Condensers, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LG-4670-104
14202-446EA 97.12 USD InStock
14202-446
SP Wilmad-LabGlass Air Condensers, SP Industries
Condensers
These borosilicate glass air condensers consist of a plain 13 mm tube with a Standard Taper 24/40 inner joint with drip tip.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty