Would you like to log in again?
SP Wilmad-LabGlass Reducing Adapters, Bushing Style, SP Industries
Supplier: SP Industries
LG-1240-118 LG-1240-126 LG-1240-120
14202-434EA 123.95 USD InStock
14202-434 14202-760 14202-970
SP Wilmad-LabGlass Reducing Adapters, Bushing Style, SP Industries
Adapters Adapters, Conical Ground Joints
This reducing adapter provides compact Standard Taper assemblies.
  • Larger [ST] size on outside bottom
  • Smaller [ST] size on top
The larger Standard Taper size is on the outside bottom of bushing while the smaller Standard Taper dimension is on top within the bore of bushing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty