Would you like to log in again?
SP Wilmad-LabGlass Capillary Melting Point Tubes, SP Industries
Supplier: SP Industries
LG-9060-106 LG-9060-100 LG-9060-102
14212-324EA 48.25 USD InStock
14212-324 14203-298 14212-392
SP Wilmad-LabGlass Capillary Melting Point Tubes, SP Industries
Tubes Melting Point Tubes and Capillaries
Fabricated from low extractable borosilicate glass, these melting point capillary tubes will not react with organic compounds.
Capillary tubes are closed at one end and packed in vials containing approximately 100 tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty