Would you like to log in again?
SP Wilmad-LabGlass All-Glass Filter Apparatus, SP Industries
Supplier: SP Industries
BP-1825-000 BP-1825-004 BP-1826-000 BP-1825-002 BP-1828-000 BP-1781-092 BP-1825-090 BP-1781-000 BP-1827-000 BP-1827-004 BP-1826-090 BP-1771-000
14204-652EA 387.39 USD InStock
14204-652 14204-740 14209-410 14210-498 14203-422 14204-504 14207-958 14206-258 14205-742 14209-536 14204-760 14207-686
SP Wilmad-LabGlass All-Glass Filter Apparatus, SP Industries
Funnels Filtration Funnels Filtration Apparatus Glass
Choose from 47mm or 90mm microfiltration FILTERWARE™ all-borosilicate glass filter apparatus consisting of a graduated funnel, a Standard Taper 40/35 glass joint connecting the coarse fritted filter support base to the filter flask, and an aluminum clamp (BP-1753).
  • Use for HPLC mobile phase filtration, analysis of particulate or bacterial contamination, or for general laboratory microfiltration
Note: Components of 47mm apparatus are interchangeable with the corresponding components of other manufacturer's items and can be used with filtering manifolds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty