Would you like to log in again?
SP Wilmad-LabGlass Filterware™ Filter Flask, SP Industries
Supplier: SP Industries
BP-1827-002
14203-552EA 412.71 USD InStock
14203-552
SP Wilmad-LabGlass Filterware™ Filter Flask, SP Industries
Flasks Filter Flasks
These microfiltration components are designed for the filtration of aqueous liquids
  • HPLC mobile phase filtration
  • Suitable for general laboratory microfiltration
The components are interchangeable with competitive product offerings and can be used with filtering manifolds.
They may be used for analysis of particulate or bacterial contamination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty