Would you like to log in again?
SP Wilmad-LabGlass Precision Grade Graduated Cylinders with Pour-Out, SP Industries
Supplier: SP Industries
LG-5520-110 LG-5520-106 LG-5520-112 LG-5520-102 LG-5520-100 LG-5520-108 LG-5520-104
14209-652EA 656.01 USD InStock
14209-652 14203-674 14202-898 14210-392 14203-672 14207-424 14209-028
SP Wilmad-LabGlass Precision Grade Graduated Cylinders with Pour-Out, SP Industries
Cylinders Measuring Cylinders
Having a great level of precision and conforming to ASTM E1272 Style 1 Class A, these borosilicate glass graduated cylinders feature a reinforced bead and pouring lip.
Calibrated “to deliver” at 20 °C and suitable for general laboratory use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty