Would you like to log in again?
SP Wilmad-LabGlass Short Path Distilling Heads with Vigreux Indents, SP Industries
Supplier: SP Industries
LG-6316-106 LG-6316-104
14203-822EA 569.76 USD InStock
14203-822 14203-826
SP Wilmad-LabGlass Short Path Distilling Heads with Vigreux Indents, SP Industries
Distillation Components Distillation Heads
This short path distillation head features Vigreux indentations in the bottom portion of the stem to improve efficiency.
  • Borosilicate glass
It is used where small quantities must be kept to a minimum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty