Would you like to log in again?
SP Wilmad-LabGlass Four-Neck Reaction Vessel Covers, SP Industries
Supplier: SP Industries
LG-8031-100 LG-8031-102
14203-970EA 814.87 USD InStock
14203-970 14204-562
SP Wilmad-LabGlass Four-Neck Reaction Vessel Covers, SP Industries
Lids Reaction Vessel Lids
Featuring a center neck and three side necks with full length outer Standard Taper joints that are 120° apart, these borosilicate glass reaction vessel lids have a conical flange
Bottom flange ID is 4" and ground for a leak-proof fit with a reaction flask. For use with reaction vessels LG-8010 through LG-8026
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty