Would you like to log in again?
SP Wilmad-LabGlass Cool Coil Condensers, SP Industries
Supplier: SP Industries
LG-5037-104 LG-5037-100 LG-5037-106
14204-424EA 866.76 USD InStock
14204-424 14204-304 14208-258
SP Wilmad-LabGlass Cool Coil Condensers, SP Industries
Condensers
Coolant is circulated through a coil in the jacket and vapors rise within the jacket itself in this condenser.
  • Borosilicate glass
Top outer and bottom inner drip tip have the same size Standard Taper joints.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty