Would you like to log in again?
SP Wilmad-LabGlass Allihn Condensers, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LG-4680-130 LG-4680-102 LG-4680-106 LG-4680-104 LG-4680-100
14204-860EA 423.41 USD InStock
14204-860 14204-346 14204-218 14206-490 14204-648
SP Wilmad-LabGlass Allihn Condensers, SP Industries
Condensers
These borosilicate glass Allihn reflux condensers have outer Standard Taper joint at top and an inner drip Standard Taper joint of the same size at bottom.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty