Would you like to log in again?
SP Wilmad-LabGlass Chromatography Columns with Fritted Disc, SP Industries
Supplier: SP Industries
LG-4567T-130 LG-4567T-106 LG-4567T-104 LG-4567T-122 LG-4567T-126
14202-702EA 237.78 USD InStock
14202-702 14204-344 14202-704 14204-566 14209-680
SP Wilmad-LabGlass Chromatography Columns with Fritted Disc, SP Industries
Chromatography Columns
These borosilicate glass plain chromatography columns have a coarse porosity fritted disc sealed into the column just above the straight bore stopcock with PTFE plug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty