Would you like to log in again?
SP Wilmad-LabGlass Cylindrical Reaction Vessels, O-Ring Flange, SP Industries
Supplier: SP Industries
LG-8071-100 LG-8075-104 LG-8075-102 LG-8071-102 LG-8075-100
14203-984EA 498.23 USD InStock
14203-984 14204-874 14204-802 14204-050 14204-790
SP Wilmad-LabGlass Cylindrical Reaction Vessels, O-Ring Flange, SP Industries
Reaction Vessels Reaction Vessels, Cylindrical Shape
These borosilicate glass cylindrical reaction vessels have a tooled flat flange with an O-ring groove for a greaseless seal
Suitable for vacuum operation. Wall thickness is uniform throughout flask. Note: Use LG-7316 clamps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty