Would you like to log in again?
SP Wilmad-LabGlass Bulb Oil Centrifuge Tubes, SP Industries
Supplier: SP Industries
LG-4492-100 LG-4490-100
14209-542EA 378.37 USD InStock
14209-542 14204-962
SP Wilmad-LabGlass Bulb Oil Centrifuge Tubes, SP Industries
Tubes Centrifuge Tubes
Calibrated 'to contain' (TC), these Goetz-type borosilicate glass bulb oil centrifuge tubes are pear shaped, similar to LG-4420, but they have a lower stem calibrated to hold 1.5 ml.
  • Reference No. ASTM D 96
Certifications: Specified in ASTM Method D 96 as suitable for a number of tests on petroleum products.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty