Would you like to log in again?
SP Wilmad-LabGlass Carboys with Hose Connection, SP Industries
Supplier: SP Industries
LG-3465-102 LG-3465-106 LG-3465-104 LG-3465-100
14205-194EA 984.21 USD InStock
14205-194 14206-110 14207-556 14208-302
SP Wilmad-LabGlass Carboys with Hose Connection, SP Industries
Bottles Carboys
Large capacity borosilicate glass bottles, offered in four different sizes, feature a hose connector sealed at neck (¹/₂ or ³/₈" connection).
All bottle necks are compatible with a #12 stopper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty