Would you like to log in again?
SP Wilmad-LabGlass Jacketed Cylindrical Reaction Vessels with Flat Flange, SP Industries
Supplier: SP Industries
LG-8089-102 LG-8089-108 LG-8089-106 LG-8089-100 LG-8089-104
14205-272EA 1599.57 USD InStock
14205-272 14207-924 14208-176 14204-628 14206-122
SP Wilmad-LabGlass Jacketed Cylindrical Reaction Vessels with Flat Flange, SP Industries
Reaction Vessels Reaction Vessels, Cylindrical Shape
These jacketed, cylindrical borosilicate glass reaction vessels are without constriction at the top and have a flat bottom.
  • Sturdy walls
  • Tooled flat flange
They feature sturdy walls and a tooled flat flange with a surface ground to assure a tight seal. There is a gently sloping contour at junction of side wall and bottom, thus preventing any weakening that might occur with a sharp shoulder. Jacket has 1/2" hose connectors.
Ordering information: Uses LG-7316 clamps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty