Would you like to log in again?
SP Wilmad-LabGlass Laboratory Grade Automatic Burettes with PTFE Plug, SP Industries
Supplier: SP Industries
LG-4241T-100 LG-4241T-104 LG-4241T-102
14203-812EA 700.38 USD InStock
14203-812 14208-260 14205-414
SP Wilmad-LabGlass Laboratory Grade Automatic Burettes with PTFE Plug, SP Industries
Burets
This inexpensive portable titration set features a 2mm straight stopcock with PTFE plug
The borosilicate glass burette is filled by pressure supplied by the rubber bulb. A small hole in the filling tube at the "O" line permits setting of the liquid level and excess titrant is returned to the bottle. Complete Set furnished with burette, bottle, rubber hose, rubber stopper, and rubber bulb.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty