Would you like to log in again?
SP Wilmad-LabGlass Cylindrical Jacketed/Indented Reaction Vessels, O-Ring Flange, SP Industries
Supplier: SP Industries
LG-8079B-106 LG-8079B-102 LG-8079B-108 LG-8079B-104
14206-260EA 1365.78 USD InStock
14206-260 14205-270 14207-786 14205-354
SP Wilmad-LabGlass Cylindrical Jacketed/Indented Reaction Vessels, O-Ring Flange, SP Industries
Reaction Vessels Reaction Vessels, Cylindrical Shape
These borosilicate glass reaction vessels are jacketed with 1/2" hose connectors.
They have four indentations for improved stirring and a tooled flat flange with an O-ring groove for a greaseless seal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty