Would you like to log in again?
SP Wilmad-LabGlass Bend Adapters with Hose Connection and Standard Taper Joint, SP Industries
Supplier: SP Industries
LG-1702-102
14206-312EA 51.11 USD InStock
14206-312
SP Wilmad-LabGlass Bend Adapters with Hose Connection and Standard Taper Joint, SP Industries
Adapters Adapters, Joints with Tube Connectors
These borosilicate glass adapters have a 90° bend to hose connector at top and a standard taper inner joint at bottom.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty