Would you like to log in again?
SP Wilmad-LabGlass Round Bottom Four-Neck Flasks, SP Industries
Supplier: SP Industries
LG-7370-204 LG-7370-278 LG-7370-178 LG-7370-172 LG-7370-212 LG-7370-188 LG-7370-270 LG-7370-196 LG-7370-194
14206-262EA 1274.24 USD InStock
14206-262 14208-504 14205-590 14207-718 14207-332 14205-444 14204-808 14206-256 14204-584
SP Wilmad-LabGlass Round Bottom Four-Neck Flasks, SP Industries
Flasks Round Bottom Flasks
Available in various sizes, these borosilicate glass round bottom flasks have four vertical necks - one center neck and three side necks which are 90° apart
All necks have Standard Taper outer joints.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty