Would you like to log in again?
SP Wilmad-LabGlass Kohlrausch Volumetric Flasks, Class A, SP Industries
Supplier: SP Industries
LG-8192-100 LG-8192-102
14207-394EA 164.97 USD InStock
14207-394 14202-462
SP Wilmad-LabGlass Kohlrausch Volumetric Flasks, Class A, SP Industries
Flasks Volumetric Flasks
Calibrated ""to contain"" at 20°C, the upper neck of these borosilicate glass flasks are enlarged to permit easy introduction of solids
Each flask is provided with a marking spot in addition to a serial number.
Ordering information: Supplied with Certificate of Compliance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty