Would you like to log in again?
SP Wilmad-LabGlass Cylindrical Reaction Vessels with Flat Flange, SP Industries
Supplier: SP Industries
LG-8080-100 LG-8080-102
14207-656EA 406.7 USD InStock
14207-656 14207-638
SP Wilmad-LabGlass Cylindrical Reaction Vessels with Flat Flange, SP Industries
Reaction Vessels Reaction Vessels, Cylindrical Shape
These borosilicate glass cylindrical reaction vessels are without constriction at the top and have a flat bottom.
  • Sturdy walls
  • Tooled flat flange
They feature sturdy walls and a tooled flat flange with a surface ground to assure a tight seal. There is a gently sloping contour at junction of side wall and bottom, thus preventing any weakening that might occur with a sharp shoulder.
Ordering information: Uses LG-7316-106 clamp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty