Would you like to log in again?
SP Wilmad-LabGlass Cylindrical Jacketed Reaction Vessels with O-Ring Flange, SP Industries
Supplier: SP Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LG-8079C-100 LG-8079C-104 LG-8079C-102
14207-900EA 1599.05 USD InStock
14207-900 14204-796 14208-676
SP Wilmad-LabGlass Cylindrical Jacketed Reaction Vessels with O-Ring Flange, SP Industries
Reaction Vessels Reaction Vessels, Cylindrical Shape
These cylindrical borosilicate glass reaction vessels are jacketed with 1/2" hose connections
They have a tooled flat flange with an O-ring groove for a greaseless seal. Suitable for vacuum operation. Wall thickness is uniform throughout flask.
Ordering information: Use LG-7316 clamps.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty